Assay Details
Target Gene Details
Entrez Gene ID: | 64137 |
Gene Name: | ATP binding cassette subfamily G member 4 |
Gene Aliases: |
WHITE2 |
Location: |
Chr.11:119149040-119162666 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABCG4 | NM_001142505.1 | NP_001135977.1 | ||
NM_022169.4 | NP_071452.2 | |||
XM_017018121.1 | XP_016873610.1 | |||
AJ300465.1 | CAC17140.1 | |||
AJ308237.1 | CAC87131.1 | |||
AK289830.1 | ||||
AK290035.1 | ||||
BC041091.1 | AAH41091.1 | |||
DA093690.1 | ||||
DA241043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv556466 | Chr.11:119106009 - 119188695 on Build GRCh38 | Loss | HINFP PDZD3 C2CD2L NLRX1 ABCG4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |