Assay Details
Target Gene Details
Entrez Gene ID: | 2132 |
Gene Name: | exostosin glycosyltransferase 2 |
Gene Aliases: |
SOTV, SSMS |
Location: |
Chr.11:44095549-44245430 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXT2 | NM_000401.3 | NP_000392.3 | ||
NM_001178083.1 | NP_001171554.1 | |||
NM_207122.1 | NP_997005.1 | |||
XM_011519950.1 | XP_011518252.1 | |||
XM_011519951.1 | XP_011518253.1 | |||
AK296713.1 | ||||
AK312375.1 | ||||
BC010058.1 | AAH10058.1 | |||
BC068545.1 | AAH68545.1 | |||
BX648142.1 | ||||
U62740.1 | AAB07008.1 | |||
U64511.1 | AAC50764.1 | |||
U72263.1 | AAB62718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832139 | Chr.11:44063681 - 44277647 on Build GRCh38 | Loss | EXT2 ACCS ALX4 |
nsv1043523 | Chr.11:44082672 - 44137619 on Build GRCh38 | Gain | EXT2 ACCS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |