Assay Details
Target Gene Details
Entrez Gene ID: | 57410 |
Gene Name: | SCY1 like pseudokinase 1 |
Gene Aliases: |
GKLP, HT019, NKTL, NTKL, P105, SCAR21, TAPK, TEIF, TRAP |
Location: |
Chr.11:65525065-65538711 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCYL1 | NM_001048218.1 | NP_001041683.1 | ||
NM_020680.3 | NP_065731.3 | |||
XM_005274118.3 | XP_005274175.1 | |||
XM_005274120.3 | XP_005274177.1 | |||
XM_005274121.3 | XP_005274178.1 | |||
XM_017018029.1 | XP_016873518.1 | |||
XM_017018030.1 | XP_016873519.1 | |||
XM_017018031.1 | XP_016873520.1 | |||
XM_017018032.1 | XP_016873521.1 | |||
XM_017018033.1 | XP_016873522.1 | |||
AB047077.1 | BAB55454.1 | |||
AB051427.1 | BAB55458.1 | |||
AB051428.1 | BAB55459.1 | |||
AF297709.1 | AAG17902.1 | |||
AK302198.1 | ||||
AK314945.1 | ||||
BC069233.1 | AAH69233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv361 | Chr.11:65520835 - 65534840 on Build GRCh38 | Insertion |
![]() |
nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)