Assay Details
Target Gene Details
Entrez Gene ID: | 55742 |
Gene Name: | parvin alpha |
Gene Aliases: |
CH-ILKBP, MXRA2 |
Location: |
Chr.11:12376441-12535356 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PARVA | NM_018222.4 | NP_060692.2 | ||
XM_005253015.3 | XP_005253072.1 | |||
AF237771.1 | AAG27173.1 | |||
AF325830.1 | AAK49911.1 | |||
AK001655.1 | BAA91815.1 | |||
AK022316.1 | BAB14009.1 | |||
AK098504.1 | ||||
BC014535.1 | AAH14535.1 | |||
BC016713.1 | AAH16713.1 | |||
DC344538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625390 | Chr.11:12320996 - 12405629 on Build GRCh38 | Gain | PARVA MICALCL |
esv3625391 | Chr.11:12333021 - 12387704 on Build GRCh38 | Gain | PARVA MICALCL |
nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss | PARVA CTR9 DKK3 MICAL2 LOC101928053 ZBED5 USP47 LOC105376545 MICALCL MIR4299 EIF4G2 SNORD97 MIR6124 MIR8070 ZBED5-AS1 LOC105376554 CSNK2A3 TEAD1 GALNT18 MRVI1 |
esv3579373 | Chr.11:12379317 - 12393635 on Build GRCh38 | Loss | PARVA |
dgv1059n100 | Chr.11:12197656 - 12685051 on Build GRCh38 | Gain | PARVA MICAL2 TEAD1 MICALCL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |