Assay Details
Target Gene Details
Entrez Gene ID: | 100289388 |
Gene Name: | KCTD21 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:78139793-78173977 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCTD21-AS1 | NR_102280.1 | |||
NR_102281.1 | ||||
DA156609.1 | ||||
DA737651.1 | ||||
DA814941.1 | ||||
DB082989.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain | AAMDC NDUFC2-KCTD14 ALG8 USP35 INTS4 RSF1 NDUFC2 KCTD21 GAB2 THRSP KCTD21-AS1 KCTD14 |
nsv516503 | Chr.11:78145131 - 78209884 on Build GRCh38 | Loss | USP35 KCTD21 KCTD21-AS1 |
nsv1047928 | Chr.11:78114229 - 78166323 on Build GRCh38 | Gain | ALG8 KCTD21-AS1 |