Assay Details
Target Gene Details
Entrez Gene ID: | 572 |
Gene Name: | BCL2 associated agonist of cell death |
Gene Aliases: |
BBC2, BCL2L8 |
Location: |
Chr.11:64269828-64284704 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BAD | NM_004322.3 | NP_004313.1 | ||
NM_032989.2 | NP_116784.1 | |||
AB451254.1 | ||||
AB451378.1 | ||||
AF021792.1 | AAB72092.1 | |||
AF031523.1 | AAB88124.1 | |||
AK291863.1 | ||||
AK303051.1 | ||||
AK309150.1 | 4 | 805 | ||
BC001901.2 | AAH01901.1 | |||
BC095431.1 | AAH95431.1 | |||
BE255791.1 | ||||
BG435829.1 | ||||
BG748336.1 | ||||
BT006678.1 | AAP35324.1 | |||
CR541935.1 | CAG46733.1 | |||
CR541959.1 | CAG46757.1 | |||
U66879.1 | AAB36516.1 |
Target Gene Details
Entrez Gene ID: | 56834 |
Gene Name: | G protein-coupled receptor 137 |
Gene Aliases: |
C11orf4, GPR137A, TM7SF1L1 |
Location: |
Chr.11:64270337-64289500 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR137 | XM_011545168.2 | XP_011543470.1 | ||
XM_011545170.2 | XP_011543472.1 | |||
XM_011545171.2 | XP_011543473.1 | |||
XM_011545172.2 | XP_011543474.1 | |||
XM_017018016.1 | XP_016873505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | GPR137 MACROD1 FKBP2 LOC105369340 DNAJC4 VEGFB PLCB3 BAD STIP1 TRPT1 FERMT3 PPP1R14B NUDT22 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | GPR137 MACROD1 FKBP2 LOC105369340 PRDX5 VEGFB MIR7155 TEX40 PLCB3 CCDC88B COX8A KCNK4-TEX40 PPP1R14B NUDT22 OTUB1 ESRRA DNAJC4 KCNK4 TRMT112 BAD STIP1 TRPT1 FERMT3 FLRT1 NAA40 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | GPR137 FKBP2 ESRRA LOC105369340 DNAJC4 PRDX5 VEGFB KCNK4 TRMT112 TEX40 PLCB3 BAD STIP1 TRPT1 FERMT3 KCNK4-TEX40 PPP1R14B NUDT22 |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | MIR1237 GPR137 FKBP2 ESRRA LOC105369340 DNAJC4 PRDX5 VEGFB MIR7155 KCNK4 TRMT112 TEX40 PLCB3 CCDC88B RPS6KA4 BAD TRPT1 FERMT3 KCNK4-TEX40 PPP1R14B NUDT22 |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | GPR137 MACROD1 FKBP2 LOC105369340 DNAJC4 VEGFB PLCB3 BAD STIP1 TRPT1 FERMT3 PPP1R14B NUDT22 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | GPR137 MACROD1 FKBP2 ESRRA LOC105369340 DNAJC4 PRDX5 VEGFB KCNK4 TRMT112 TEX40 PLCB3 BAD STIP1 TRPT1 FERMT3 KCNK4-TEX40 PPP1R14B NUDT22 FLRT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |