Assay Details
Target Gene Details
Entrez Gene ID: | 80313 |
Gene Name: | leucine rich repeat containing 27 |
Gene Aliases: |
- |
Location: |
Chr.10:132330063-132381506 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054569 | Chr.10:132009350 - 132351508 on Build GRCh38 | Gain | JAKMIP3 STK32C LRRC27 DPYSL4 |
esv3624988 | Chr.10:132338306 - 132366249 on Build GRCh38 | Gain | LRRC27 |
nsv1051555 | Chr.10:132295188 - 132415461 on Build GRCh38 | Gain | PWWP2B STK32C LRRC27 |
esv3624986 | Chr.10:132322882 - 132551350 on Build GRCh38 | Gain | PWWP2B STK32C C10orf91 LOC107984282 LRRC27 INPP5A LOC105378568 |
esv2761642 | Chr.10:132048518 - 132365985 on Build GRCh38 | Gain | JAKMIP3 STK32C LRRC27 DPYSL4 |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain | JAKMIP3 MIR378C LINC01164 ADGRA1 LINC01168 LINC01167 BNIP3 C10orf91 LOC107984282 ADGRA1-AS1 DPYSL4 INPP5A PWWP2B NKX6-2 CFAP46 LINC01166 TCERG1L STK32C LRRC27 LOC105378568 TCERG1L-AS1 PPP2R2D |
nsv1052405 | Chr.10:132336481 - 132390781 on Build GRCh38 | Gain | LRRC27 |