Assay Details
Target Gene Details
Entrez Gene ID: | 22986 |
Gene Name: | sortilin related VPS10 domain containing receptor 3 |
Gene Aliases: |
SORCS |
Location: |
Chr.10:104641101-105265242 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SORCS3 | NM_014978.2 | NP_055793.1 | ||
AB028982.1 | BAA83011.2 | |||
BC117417.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3579100 | Chr.10:104573121 - 104983037 on Build GRCh38 | Loss | SORCS3 SORCS3-AS1 |
nsv831972 | Chr.10:104523290 - 104689830 on Build GRCh38 | Gain | SORCS3 SORCS3-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |