Assay Details
Target Gene Details
Entrez Gene ID: | 3688 |
Gene Name: | integrin subunit beta 1 |
Gene Aliases: |
CD29, FNRB, GPIIA, MDF2, MSK12, VLA-BETA, VLAB |
Location: |
Chr.10:32900318-32958365 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGB1 | NM_002211.3 | NP_002202.2 | ||
NM_033668.2 | NP_391988.1 | |||
NM_133376.2 | NP_596867.1 | |||
AK291697.1 | ||||
BC020057.1 | AAH20057.1 | |||
BX537407.1 | CAD97649.1 | |||
CX868374.1 | ||||
CX871886.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831827 | Chr.10:32817738 - 32985904 on Build GRCh38 | Loss | CCDC7 SNORA86 ITGB1 |
nsv550383 | Chr.10:32898536 - 32960205 on Build GRCh38 | Loss | SNORA86 ITGB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |