Assay Details
Target Gene Details
Entrez Gene ID: | 83938 |
Gene Name: | chromosome 10 open reading frame 11 |
Gene Aliases: |
CDA017 |
Location: |
Chr.10:75431646-76559220 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C10orf11 | NM_001305581.1 | NP_001292510.1 | ||
NR_131178.1 | ||||
XM_011540256.2 | XP_011538558.1 | |||
XM_011540257.2 | XP_011538559.1 | |||
XM_017016759.1 | XP_016872248.1 | |||
XM_017016760.1 | XP_016872249.1 | |||
XM_017016761.1 | XP_016872250.1 | |||
BM720401.1 | ||||
BX105111.1 | ||||
CR933639.1 | ||||
HY006640.1 |
Target Gene Details
Entrez Gene ID: | 105378367 |
Gene Name: | uncharacterized LOC105378367 |
Gene Aliases: |
- |
Location: |
Chr.10:75712455-75743848 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105378367 | NR_132791.1 | |||
HY007587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831914 | Chr.10:75590441 - 75760988 on Build GRCh38 | Gain | LOC105378367 C10orf11 |
esv2759768 | Chr.10:75475179 - 76010691 on Build GRCh38 | Gain+Loss | LOC105378367 MIR606 C10orf11 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |