Assay Details
Target Gene Details
Entrez Gene ID: | 6387 |
Gene Name: | C-X-C motif chemokine ligand 12 |
Gene Aliases: |
IRH, PBSF, SCYB12, SDF1, TLSF, TPAR1 |
Location: |
Chr.10:44292088-44385097 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CXCL12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508579 | Chr.10:44214322 - 44303808 on Build GRCh38 | Deletion | C10orf142 CXCL12 |
esv3352463 | Chr.10:44294998 - 44295996 on Build GRCh38 | Duplication | CXCL12 |
nsv483001 | Chr.10:44208843 - 44393301 on Build GRCh38 | Loss | C10orf142 CXCL12 |
nsv1038511 | Chr.10:44101437 - 44374582 on Build GRCh38 | Loss | C10orf142 CXCL12 |