Assay Details
Target Gene Details
Entrez Gene ID: | 102724323 |
Gene Name: | uncharacterized LOC102724323 |
Gene Aliases: |
- |
Location: |
Chr.10:45444568-45453121 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC102724323 | NR_120674.1 | |||
AI671881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1043536 | Chr.10:45420493 - 45580787 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3891797 | Chr.10:45008026 - 45674466 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)