Assay Details
Target Gene Details
Entrez Gene ID: | 9317 |
Gene Name: | phosphotriesterase related |
Gene Aliases: |
HPHRP, RPR-1 |
Location: |
Chr.10:16436943-16517962 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTER | NM_001001484.2 | NP_001001484.1 | ||
NM_001261836.1 | NP_001248765.1 | |||
NM_001261837.1 | NP_001248766.1 | |||
NM_001261838.1 | NP_001248767.1 | |||
NM_030664.4 | NP_109589.2 | |||
XM_017016928.1 | XP_016872417.1 | |||
XM_017016929.1 | XP_016872418.1 | |||
XM_017016930.1 | XP_016872419.1 | |||
AF212237.1 | AAK14923.1 | |||
AK095486.1 | ||||
AK314910.1 | ||||
BC015092.1 | AAH15092.1 | |||
BC050411.1 | AAH50411.1 | |||
BU172463.1 | ||||
BX648556.1 | ||||
DA146922.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv550019 | Chr.10:16463718 - 16478785 on Build GRCh38 | Loss | PTER |
nsv1046833 | Chr.10:16385108 - 16530259 on Build GRCh38 | Gain | C1QL3 PTER |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |