Assay Details
Target Gene Details
Entrez Gene ID: | 22921 |
Gene Name: | methionine sulfoxide reductase B2 |
Gene Aliases: |
CBS-1, CBS1, CGI-131, MSRB, PILB |
Location: |
Chr.10:23095498-23122013 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MSRB2 | NM_012228.3 | NP_036360.3 | ||
XM_011519426.2 | XP_011517728.1 | |||
AF122004.1 | AAD38899.1 | |||
AF151889.1 | AAD34126.1 | |||
BC018030.1 | AAH18030.1 | |||
BC117471.1 | ||||
BC130380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1161931 | Chr.10:23104297 - 23109157 on Build GRCh38 | Deletion | MSRB2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |