Assay Details
Target Gene Details
Entrez Gene ID: | 114815 |
Gene Name: | sortilin related VPS10 domain containing receptor 1 |
Gene Aliases: |
hSorCS |
Location: |
Chr.10:106573663-107181138 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SORCS1 | NM_001013031.2 | NP_001013049.1 | ||
NM_001206569.1 | NP_001193498.1 | |||
NM_001206570.1 | NP_001193499.1 | |||
NM_001206571.1 | NP_001193500.1 | |||
NM_001206572.1 | NP_001193501.1 | |||
NM_052918.4 | NP_443150.3 | |||
XM_011539199.2 | XP_011537501.1 | |||
XM_017015614.1 | XP_016871103.1 | |||
XM_017015615.1 | XP_016871104.1 | |||
XM_017015616.1 | XP_016871105.1 | |||
XM_017015617.1 | XP_016871106.1 | |||
XM_017015618.1 | XP_016871107.1 | |||
AB209142.1 | BAD92379.1 | |||
AF284756.1 | AAL56667.1 | |||
AK074581.1 | ||||
AK123752.1 | ||||
AK125464.1 | ||||
AK126905.1 | ||||
AK289797.1 | ||||
AY099452.1 | AAM43811.1 | |||
AY099453.1 | AAM43812.1 | |||
BC131597.1 | ||||
EU281343.1 | ||||
EU281344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831977 | Chr.10:106466255 - 106653420 on Build GRCh38 | Gain | SORCS1 |
nsv1047446 | Chr.10:105797401 - 106616713 on Build GRCh38 | Gain | SORCS1 LOC101927549 LOC105378470 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |