Assay Details
Target Gene Details
Entrez Gene ID: | 196051 |
Gene Name: | phospholipid phosphatase 4 |
Gene Aliases: |
DPPL2, PPAPDC1, PPAPDC1A |
Location: |
Chr.10:120456954-120600123 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLPP4 | NM_001030059.2 | NP_001025230.1 | ||
NM_001318166.1 | NP_001305095.1 | |||
NM_001318167.1 | NP_001305096.1 | |||
NM_001318168.1 | NP_001305097.1 | |||
NM_001318169.1 | NP_001305098.1 | |||
NR_134516.1 | ||||
XM_005269592.2 | XP_005269649.1 | |||
XM_006717685.3 | XP_006717748.1 | |||
XM_006717686.3 | XP_006717749.1 | |||
XM_011539444.2 | XP_011537746.1 | |||
XM_011539445.2 | XP_011537747.1 | |||
XM_017015820.1 | XP_016871309.1 | |||
XM_017015823.1 | XP_016871312.1 | |||
XM_017015824.1 | XP_016871313.1 | |||
AK296273.1 | ||||
AK310178.1 | ||||
BC101267.2 | ||||
BC101268.2 | ||||
BC101269.1 | AAI01270.1 | |||
BC122535.1 | ||||
BC132787.1 | ||||
BI550027.1 | ||||
DB473853.2 | ||||
DB495310.2 | ||||
DR000933.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |