Assay Details
Target Gene Details
Entrez Gene ID: | 22931 |
Gene Name: | RAB18, member RAS oncogene family |
Gene Aliases: |
RAB18LI1, WARBM3 |
Location: |
Chr.10:27504174-27542237 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB18 | NM_001256410.1 | NP_001243339.1 | ||
NM_001256411.1 | NP_001243340.1 | |||
NM_001256412.1 | NP_001243341.1 | |||
NM_001256415.1 | NP_001243344.1 | |||
NM_021252.4 | NP_067075.1 | |||
NR_046172.1 | ||||
AF087860.1 | AAP97170.1 | |||
AF136974.1 | AAG49435.1 | |||
AF137372.1 | AAF61433.1 | |||
AF498950.1 | AAM21098.1 | |||
AK001555.1 | ||||
AK223153.1 | BAD96873.1 | |||
AK295443.1 | ||||
AK297629.1 | ||||
AK299457.1 | ||||
AL136734.1 | CAB66668.1 | |||
AY574034.1 | AAU08232.1 | |||
BC015014.2 | AAH15014.1 | |||
BC029350.1 | AAH29350.1 | |||
BP210053.1 | ||||
BT009840.1 | AAP88842.1 | |||
CN357871.1 | ||||
CR533455.1 | CAG38486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1045257 | Chr.10:27283511 - 27638017 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv550241 | Chr.10:27280592 - 27736842 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv8614 | Chr.10:27215558 - 27539477 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3622708 | Chr.10:27138085 - 27696800 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2734606 | Chr.10:27447765 - 29432048 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)