Assay Details
Target Gene Details
Entrez Gene ID: | 80313 |
Gene Name: | leucine rich repeat containing 27 |
Gene Aliases: |
- |
Location: |
Chr.10:132330063-132381506 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC27 | NM_001143757.1 | NP_001137229.1 | ||
NM_001143758.1 | NP_001137230.1 | |||
NM_001143759.1 | NP_001137231.1 | |||
NM_001309474.1 | NP_001296403.1 | |||
NM_030626.2 | NP_085129.1 | |||
XM_006717983.2 | XP_006718046.1 | |||
XM_006717984.2 | XP_006718047.1 | |||
XM_011540208.1 | XP_011538510.1 | |||
XM_011540209.1 | XP_011538511.1 | |||
XM_011540210.2 | XP_011538512.1 | |||
XM_017016690.1 | XP_016872179.1 | |||
XM_017016691.1 | XP_016872180.1 | |||
XM_017016692.1 | XP_016872181.1 | |||
XM_017016693.1 | XP_016872182.1 | |||
XM_017016694.1 | XP_016872183.1 | |||
AB051461.1 | ||||
AK093250.1 | BAC04108.1 | |||
AK097418.1 | ||||
AK098652.1 | BAC05366.1 | |||
AK295953.1 | ||||
AK301422.1 | ||||
BC050097.1 | AAH50097.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054569 | Chr.10:132009350 - 132351508 on Build GRCh38 | Gain | LRRC27 JAKMIP3 STK32C DPYSL4 |
esv3624988 | Chr.10:132338306 - 132366249 on Build GRCh38 | Gain | LRRC27 |
nsv1051555 | Chr.10:132295188 - 132415461 on Build GRCh38 | Gain | LRRC27 STK32C PWWP2B |
esv3624986 | Chr.10:132322882 - 132551350 on Build GRCh38 | Gain | LOC107984282 LRRC27 LOC105378568 C10orf91 STK32C PWWP2B INPP5A |
esv2761642 | Chr.10:132048518 - 132365985 on Build GRCh38 | Gain | LRRC27 JAKMIP3 STK32C DPYSL4 |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain | NKX6-2 CFAP46 ADGRA1 JAKMIP3 LINC01167 C10orf91 STK32C DPYSL4 PPP2R2D INPP5A BNIP3 LOC107984282 LINC01168 LRRC27 LOC105378568 ADGRA1-AS1 TCERG1L LINC01164 TCERG1L-AS1 MIR378C LINC01166 PWWP2B |
nsv1052405 | Chr.10:132336481 - 132390781 on Build GRCh38 | Gain | LRRC27 |
dgv121n27 | Chr.10:132345217 - 132477022 on Build GRCh38 | Loss | LRRC27 LOC105378568 C10orf91 PWWP2B |
esv3577872 | Chr.10:132342397 - 132412909 on Build GRCh38 | Gain | LRRC27 PWWP2B |