Assay Details
Target Gene Details
Entrez Gene ID: | 288 |
Gene Name: | ankyrin 3, node of Ranvier (ankyrin G) |
Gene Aliases: |
ANKYRIN-G, MRT37 |
Location: |
Chr.10:60026298-60733526 on Build GRCh38 |
Assay Gene Location: | Within Intron 50 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv551176 | Chr.10:60042584 - 60054294 on Build GRCh38 | Loss | ANK3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |