Assay Details
Target Gene Details
Entrez Gene ID: | 22931 |
Gene Name: | RAB18, member RAS oncogene family |
Gene Aliases: |
RAB18LI1, WARBM3 |
Location: |
Chr.10:27504174-27542237 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB18 | NM_001256410.1 | 1 | 68 | NP_001243339.1 |
NM_001256411.1 | 1 | 68 | NP_001243340.1 | |
NM_001256412.1 | 1 | 68 | NP_001243341.1 | |
NM_001256415.1 | 1 | 68 | NP_001243344.1 | |
NM_021252.4 | 1 | 68 | NP_067075.1 | |
NR_046172.1 | 1 | 68 | ||
CN357871.1 | 1 | 137 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1045257 | Chr.10:27283511 - 27638017 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv3622724 | Chr.10:27412809 - 27505337 on Build GRCh38 | Gain |
![]() ![]() |
nsv550241 | Chr.10:27280592 - 27736842 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv8614 | Chr.10:27215558 - 27539477 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3622708 | Chr.10:27138085 - 27696800 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2734606 | Chr.10:27447765 - 29432048 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv825319 | Chr.10:27503878 - 27506947 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)