Assay Details
Target Gene Details
Entrez Gene ID: | 29760 |
Gene Name: | B-cell linker |
Gene Aliases: |
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca |
Location: |
Chr.10:96189171-96271576 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain | TLL2 OPALIN BLNK DNTT |
nsv831947 | Chr.10:96085409 - 96258168 on Build GRCh38 | Gain+Loss | BLNK ENTPD1-AS1 ZNF518A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |