Assay Details
Target Gene Details
Entrez Gene ID: | 1390 |
Gene Name: | cAMP responsive element modulator |
Gene Aliases: |
CREM-2, ICER, hCREM-2 |
Location: |
Chr.10:35126841-35212958 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1039051 | Chr.10:35049745 - 35203997 on Build GRCh38 | Gain | MIR3611 CREM CUL2 |
dgv707n100 | Chr.10:35130935 - 35383078 on Build GRCh38 | Gain | CCNY CREM |
dgv706n100 | Chr.10:34881487 - 35230186 on Build GRCh38 | Gain | MIR3611 CREM CUL2 |
nsv550398 | Chr.10:34885707 - 35271661 on Build GRCh38 | Gain | MIR3611 CCNY CREM CUL2 |
esv3576372 | Chr.10:35088547 - 35160753 on Build GRCh38 | Gain | CREM CUL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |