Assay Details
Target Gene Details
Entrez Gene ID: | 240 |
Gene Name: | arachidonate 5-lipoxygenase |
Gene Aliases: |
5-LO, 5-LOX, 5LPG, LOG5 |
Location: |
Chr.10:45374166-45446121 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALOX5 | NM_000698.4 | NP_000689.1 | ||
NM_001256153.2 | NP_001243082.1 | |||
NM_001256154.2 | NP_001243083.1 | |||
NM_001320861.1 | NP_001307790.1 | |||
NM_001320862.1 | NP_001307791.1 | |||
XM_017016012.1 | XP_016871501.1 | |||
AB208946.1 | BAD92183.1 | |||
AK225603.1 | ||||
AK315223.1 | ||||
BC130332.1 | ||||
BC132677.1 | ||||
BC143985.1 | ||||
HM592258.1 | ||||
HM592259.1 | ||||
HM592260.1 | ||||
HM592261.1 | ||||
HY102907.1 | ||||
J03571.1 | AAA65450.1 | |||
J03600.1 | AAA36183.1 | |||
JQ728550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3891797 | Chr.10:45008026 - 45674466 on Build GRCh38 | Gain | MARCH8 ZFAND4 RSU1P2 ALOX5 MIR3156-1 LOC102724323 ANKRD30BP3 OR13A1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |