Assay Details
Target Gene Details
Entrez Gene ID: | 23185 |
Gene Name: | La ribonucleoprotein domain family member 4B |
Gene Aliases: |
KIAA0217, LARP5 |
Location: |
Chr.10:806914-931900 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LARP4B | NM_015155.2 | NP_055970.1 | ||
XM_005252432.3 | XP_005252489.1 | |||
XM_005252435.3 | XP_005252492.1 | |||
XM_011519435.2 | XP_011517737.1 | |||
XM_017015987.1 | XP_016871476.1 | |||
XM_017015988.1 | XP_016871477.1 | |||
XM_017015989.1 | XP_016871478.1 | |||
XM_017015990.1 | XP_016871479.1 | |||
XM_017015992.1 | XP_016871481.1 | |||
XM_017015993.1 | XP_016871482.1 | |||
XM_017015994.1 | XP_016871483.1 | |||
BC131630.1 | ||||
BC152443.1 | ||||
CR749653.1 | CAH18446.1 | |||
D86971.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549753 | Chr.10:412537 - 848898 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv549766 | Chr.10:790175 - 842133 on Build GRCh38 | Loss |
![]() |
nsv1048707 | Chr.10:788499 - 869594 on Build GRCh38 | Gain |
![]() |
nsv549765 | Chr.10:688289 - 1083587 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)