Assay Details
Target Gene Details
Entrez Gene ID: | 54518 |
Gene Name: | amyloid beta precursor protein binding family B member 1 interacting protein |
Gene Aliases: |
INAG1, PREL1, RARP1, RIAM |
Location: |
Chr.10:26438324-26567879 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APBB1IP | NM_019043.3 | NP_061916.3 | ||
XM_011519514.2 | XP_011517816.1 | |||
AK000812.1 | BAA91384.1 | |||
AK303597.1 | ||||
AK308916.1 | ||||
AY152730.1 | AAN75525.1 | |||
BC035636.1 | AAH35636.1 | |||
BC054516.1 | AAH54516.1 | |||
BM150270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734362 | Chr.10:26419040 - 26459311 on Build GRCh38 | Deletion |
![]() |
nsv1043832 | Chr.10:26380111 - 26520471 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)