Assay Details
Target Gene Details
Entrez Gene ID: | 10006 |
Gene Name: | abl interactor 1 |
Gene Aliases: |
ABI-1, ABLBP4, E3B1, NAP1BP, SSH3BP, SSH3BP1 |
Location: |
Chr.10:26746596-26861087 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831812 | Chr.10:26772577 - 26945338 on Build GRCh38 | Gain | LINC00202-1 ABI1 ANKRD26 |
esv3415799 | Chr.10:26640952 - 26944612 on Build GRCh38 | Duplication | PDSS1 LINC00202-1 ABI1 LINC00202-2 ANKRD26 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |