Assay Details
Target Gene Details
Entrez Gene ID: | 51196 |
Gene Name: | phospholipase C epsilon 1 |
Gene Aliases: |
NPHS3, PLCE, PPLC |
Location: |
Chr.10:93993989-94329447 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCE1 | NM_001288989.1 | NP_001275918.1 | ||
NM_016341.3 | NP_057425.3 | |||
XM_006717885.3 | XP_006717948.1 | |||
XM_006717888.3 | XP_006717951.1 | |||
XM_006717889.3 | XP_006717952.1 | |||
XM_011539849.2 | XP_011538151.1 | |||
XM_011539851.2 | XP_011538153.1 | |||
XM_011539852.2 | XP_011538154.1 | |||
XM_017016310.1 | XP_016871799.1 | |||
XM_017016311.1 | XP_016871800.1 | |||
AB040949.2 | ||||
AF190642.2 | AAG17145.2 | |||
BC140705.1 | ||||
BC144286.1 | ||||
BC151854.1 |
Target Gene Details
Entrez Gene ID: | 101927049 |
Gene Name: | PLCE1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.10:94081950-94108794 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCE1-AS2 | NR_120616.1 | |||
AK098548.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |