Assay Details
Target Gene Details
Entrez Gene ID: | 643414 |
Gene Name: | lipase family member K |
Gene Aliases: |
LIPL2, bA186O14.2 |
Location: |
Chr.10:88706224-88753092 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPK | XM_011540073.2 | XP_011538375.1 | ||
XM_011540075.1 | XP_011538377.1 | |||
XM_011540077.1 | XP_011538379.1 | |||
XM_017016545.1 | XP_016872034.1 | |||
XM_017016546.1 | XP_016872035.1 | |||
XM_017016547.1 | XP_016872036.1 | |||
XM_017016548.1 | XP_016872037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1043015 | Chr.10:88707310 - 88833935 on Build GRCh38 | Gain | LIPM LIPK ANKRD22 LIPN |
nsv831941 | Chr.10:88650731 - 88827575 on Build GRCh38 | Gain | LIPM LIPK ANKRD22 LIPN LIPF |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |