Assay Details
Target Gene Details
Entrez Gene ID: | 55328 |
Gene Name: | renalase, FAD dependent amine oxidase |
Gene Aliases: |
C10orf59, RENALASE |
Location: |
Chr.10:88176458-88583860 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RNLS | XM_011539924.2 | XP_011538226.1 | ||
XM_017016382.1 | XP_016871871.1 | |||
AK296262.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831940 | Chr.10:88051490 - 88229442 on Build GRCh38 | Gain | RNLS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |