Assay Details
Target Gene Details
Entrez Gene ID: | 54780 |
Gene Name: | NSE4 homolog A, SMC5-SMC6 complex component |
Gene Aliases: |
C10orf86, NS4EA, NSE4A |
Location: |
Chr.10:121957088-121975228 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NSMCE4A | NM_001167865.1 | NP_001161337.1 | ||
NM_017615.2 | NP_060085.2 | |||
XM_005269929.4 | XP_005269986.1 | |||
XM_005269930.3 | XP_005269987.1 | |||
XM_011539910.2 | XP_011538212.1 | |||
XM_017016359.1 | XP_016871848.1 | |||
AF258584.1 | AAG23787.1 | |||
AK000010.1 | BAA90881.1 | |||
AK056579.1 | ||||
AK222487.1 | BAD96207.1 | |||
AK301658.1 | ||||
AK308687.1 | ||||
BC005212.1 | AAH05212.1 | |||
BC062427.1 | AAH62427.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |