Assay Details
Target Gene Details
Entrez Gene ID: | 8036 |
Gene Name: | SHOC2, leucine rich repeat scaffold protein |
Gene Aliases: |
SIAA0862, SOC2, SUR8 |
Location: |
Chr.10:110919370-111013667 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SHOC2 | NM_001269039.2 | NP_001255968.1 | ||
NM_001324336.1 | NP_001311265.1 | |||
NM_001324337.1 | NP_001311266.1 | |||
NM_007373.3 | NP_031399.2 | |||
NR_136749.1 | ||||
XM_017016702.1 | XP_016872191.1 | |||
XM_017016703.1 | XP_016872192.1 | |||
XM_017016704.1 | XP_016872193.1 | |||
AB020669.1 | ||||
AF054828.1 | AAC25698.1 | |||
AK292833.1 | ||||
AK293271.1 | ||||
BX649055.1 | ||||
DB451165.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1035602 | Chr.10:110876295 - 111021655 on Build GRCh38 | Loss | MIR4680 MIR548E SHOC2 PDCD4 RPL13AP6 BBIP1 |
esv3624545 | Chr.10:110948631 - 110987680 on Build GRCh38 | Loss | SHOC2 |
nsv831985 | Chr.10:110892331 - 111043345 on Build GRCh38 | Gain+Loss | MIR4680 MIR548E SHOC2 PDCD4 RPL13AP6 BBIP1 |
esv2674974 | Chr.10:110948807 - 110987520 on Build GRCh38 | Deletion | SHOC2 |
dgv974n100 | Chr.10:110924064 - 111035401 on Build GRCh38 | Loss | MIR548E SHOC2 RPL13AP6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |