Assay Details
Target Gene Details
Entrez Gene ID: | 143187 |
Gene Name: | vesicle transport through interaction with t-SNAREs 1A |
Gene Aliases: |
MMDS3, MVti1, VTI1RP2, Vti1-rp2 |
Location: |
Chr.10:112446988-112947204 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VTI1A | NM_001318203.1 | NP_001305132.1 | ||
NM_001318205.1 | NP_001305134.1 | |||
NM_145206.3 | NP_660207.2 | |||
XM_005269544.4 | XP_005269601.1 | |||
XM_005269545.4 | XP_005269602.1 | |||
XM_005269546.3 | XP_005269603.1 | |||
XM_005269547.4 | XP_005269604.1 | |||
XM_006717637.2 | XP_006717700.1 | |||
XM_011539328.2 | XP_011537630.1 | |||
XM_011539330.2 | XP_011537632.1 | |||
XM_011539331.2 | XP_011537633.1 | |||
XM_011539333.2 | XP_011537635.1 | |||
XM_017015745.1 | XP_016871234.1 | |||
XM_017015746.1 | XP_016871235.1 | |||
XM_017015747.1 | XP_016871236.1 | |||
XM_017015748.1 | XP_016871237.1 | |||
AK303646.1 | ||||
BC017052.1 | AAH17052.1 | |||
BI909044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831990 | Chr.10:112349267 - 112531997 on Build GRCh38 | Loss | ZDHHC6 ACSL5 GUCY2GP VTI1A |
nsv831991 | Chr.10:112413098 - 112582139 on Build GRCh38 | Loss | ZDHHC6 ACSL5 VTI1A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |