Assay Details
Target Gene Details
Entrez Gene ID: | 1979 |
Gene Name: | eukaryotic translation initiation factor 4E binding protein 2 |
Gene Aliases: |
4EBP2, PHASII |
Location: |
Chr.10:70404105-70428618 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF4EBP2 | NM_004096.4 | NP_004087.1 | ||
AK313724.1 | ||||
BC005057.1 | AAH05057.1 | |||
BC050633.1 | AAH50633.1 | |||
BG717474.1 | ||||
BT007317.1 | AAP35981.1 | |||
CR542240.1 | CAG47036.1 | |||
L36056.1 | AAA62270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss | NEUROG3 SAR1A PPA1 AIFM2 EIF4EBP2 H2AFY2 TSPAN15 C10orf35 COL13A1 PALD1 TYSND1 PRF1 TBATA NPFFR1 ADAMTS14 NODAL LRRC20 |
nsv1043245 | Chr.10:70290210 - 70915897 on Build GRCh38 | Loss | PALD1 PRF1 TBATA PCBD1 EIF4EBP2 ADAMTS14 SGPL1 NODAL LRRC20 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |