Assay Details
Target Gene Details
Entrez Gene ID: | 10659 |
Gene Name: | CUGBP, Elav-like family member 2 |
Gene Aliases: |
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR |
Location: |
Chr.10:10462515-11336675 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CELF2 | NM_001326317.1 | NP_001313246.1 | ||
NM_001326318.1 | NP_001313247.1 | |||
NM_001326319.1 | NP_001313248.1 | |||
NM_001326320.1 | NP_001313249.1 | |||
NM_001326321.1 | NP_001313250.1 | |||
NM_001326323.1 | NP_001313252.1 | |||
XM_017015557.1 | XP_016871046.1 | |||
XM_017015568.1 | XP_016871057.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3622328 | Chr.10:10479485 - 10492671 on Build GRCh38 | Gain | CELF2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |