Assay Details
Target Gene Details
Entrez Gene ID: | 64429 |
Gene Name: | zinc finger DHHC-type containing 6 |
Gene Aliases: |
DHHC-6, ZNF376 |
Location: |
Chr.10:112424411-112448335 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831990 | Chr.10:112349267 - 112531997 on Build GRCh38 | Loss | GUCY2GP ACSL5 ZDHHC6 VTI1A |
nsv831991 | Chr.10:112413098 - 112582139 on Build GRCh38 | Loss | ACSL5 ZDHHC6 VTI1A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |