Assay Details
Target Gene Details
Entrez Gene ID: | 56681 |
Gene Name: | secretion associated Ras related GTPase 1A |
Gene Aliases: |
SAR1, SARA1, Sara, masra2 |
Location: |
Chr.10:70150204-70170529 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAR1A | NM_001142648.1 | NP_001136120.1 | ||
NM_020150.4 | NP_064535.1 | |||
AF087897.1 | AAP97196.1 | |||
AF217959.1 | AAQ13891.1 | |||
AF261717.1 | AAF81741.1 | |||
AF274026.1 | AAM69363.1 | |||
AK298591.1 | ||||
AK312469.1 | ||||
AL136724.1 | CAB66658.1 | |||
AY008268.1 | AAG16638.1 | |||
BC003658.2 | AAH03658.1 | |||
BG327663.1 | ||||
DB216796.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv551479 | Chr.10:70007598 - 70179618 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)