Assay Details
Target Gene Details
Entrez Gene ID: | 84647 |
Gene Name: | phospholipase A2 group XIIB |
Gene Aliases: |
FKSG71, GXIIB, GXIIIsPLA2, PLA2G13, sPLA2-GXIIB |
Location: |
Chr.10:72934762-72955044 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G12B | NM_001318124.1 | NP_001305053.1 | ||
NM_001318125.1 | NP_001305054.1 | |||
NM_032562.4 | NP_115951.2 | |||
XM_005270244.4 | XP_005270301.1 | |||
AF339053.1 | AAL09472.1 | |||
AF349540.1 | AAK30168.1 | |||
AK311349.1 | ||||
AK313712.1 | ||||
AY358032.1 | AAQ92918.1 | |||
BC093996.1 | AAH93996.1 | |||
BC111946.1 | AAI11947.1 | |||
BC143532.1 | ||||
BP274516.1 | ||||
DA536331.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528473 | Chr.10:72911853 - 73049838 on Build GRCh38 | Gain | PLA2G12B OIT3 P4HA1 |
esv3623807 | Chr.10:72948186 - 72952432 on Build GRCh38 | Loss | PLA2G12B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |