Assay Details
Target Gene Details
Entrez Gene ID: | 53358 |
Gene Name: | SHC adaptor protein 3 |
Gene Aliases: |
N-Shc, NSHC, RAI, SHCC |
Location: |
Chr.9:89005771-89182308 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SHC3 | NM_016848.5 | NP_058544.3 | ||
XM_011518785.2 | XP_011517087.1 | |||
XM_011518786.2 | XP_011517088.1 | |||
BC026314.1 | AAH26314.1 | |||
BX641139.1 | ||||
D84361.1 | BAA12323.1 | |||
KU176125.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3621014 | Chr.9:88877891 - 89641453 on Build GRCh38 | Gain | S1PR3 SEMA4D SECISBP2 SHC3 MIR3153 UNQ6494 GADD45G C9orf47 CKS2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |