Assay Details
Target Gene Details
Entrez Gene ID: | 340485 |
Gene Name: | alkaline ceramidase 2 |
Gene Aliases: |
ALKCDase2, ASAH3L |
Location: |
Chr.9:19408927-19455173 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACER2 | NM_001010887.2 | NP_001010887.2 | ||
XM_005251448.3 | XP_005251505.1 | |||
XM_011517858.1 | XP_011516160.1 | |||
AF370405.1 | AAQ15241.1 | |||
AK123581.1 | ||||
AY312516.1 | AAQ85132.1 | |||
BC092487.1 | AAH92487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1025392 | Chr.9:19381646 - 19478974 on Build GRCh38 | Gain | ACER2 |
nsv613731 | Chr.9:19400353 - 19448475 on Build GRCh38 | Loss | ACER2 |
esv3619907 | Chr.9:19322693 - 19441414 on Build GRCh38 | Gain | ACER2 DENND4C RPS6 |
nsv613730 | Chr.9:19281101 - 19516865 on Build GRCh38 | Gain | ACER2 DENND4C SLC24A2 RPS6 |
esv3619911 | Chr.9:19379247 - 19517379 on Build GRCh38 | Gain | ACER2 SLC24A2 RPS6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |