Assay Details
Target Gene Details
Entrez Gene ID: | 2650 |
Gene Name: | glucosaminyl (N-acetyl) transferase 1, core 2 |
Gene Aliases: |
C2GNT, C2GNT-L, C2GNT1, G6NT, NACGT2, NAGCT2 |
Location: |
Chr.9:76441666-76507416 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GCNT1 | NM_001097633.1 | NP_001091102.1 | ||
NM_001097634.1 | NP_001091103.1 | |||
NM_001490.4 | NP_001481.2 | |||
XM_017014620.1 | XP_016870109.1 | |||
XM_017014621.1 | XP_016870110.1 | |||
AK313343.1 | ||||
AL832647.2 | CAD89956.1 | |||
AY196293.1 | ||||
AY196294.1 | ||||
AY196295.1 | ||||
BP359964.1 | ||||
DA479942.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |