Assay Details
Target Gene Details
Entrez Gene ID: | 7013 |
Gene Name: | telomeric repeat binding factor 1 |
Gene Aliases: |
PIN2, TRBF1, TRF, TRF1, hTRF1-AS, t-TRF1 |
Location: |
Chr.8:73008862-73047752 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TERF1 | NM_003218.3 | NP_003209.2 | ||
NM_017489.2 | NP_059523.2 | |||
XM_005251291.3 | XP_005251348.1 | |||
XM_005251292.3 | XP_005251349.1 | |||
XM_011517582.2 | XP_011515884.1 | |||
XM_017013792.1 | XP_016869281.1 | |||
AF003001.1 | AAB81137.1 | |||
AK292529.1 | ||||
AY576778.1 | AAT81150.1 | |||
BC029378.1 | AAH29378.1 | |||
U40705.1 | AAB54036.1 | |||
U74382.1 | AAB53363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831354 | Chr.8:72906156 - 73065096 on Build GRCh38 | Gain | SBSPON KCNB2 TERF1 |
esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion | TRPA1 RDH10 C8orf89 RDH10-AS1 STAU2-AS1 LOC392232 TERF1 MSC RPL7 STAU2 SBSPON MSC-AS1 KCNB2 LOC101926908 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |