Assay Details
Target Gene Details
Entrez Gene ID: | 10404 |
Gene Name: | carboxypeptidase Q |
Gene Aliases: |
LDP, PGCP |
Location: |
Chr.8:96645227-97143503 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CPQ | NM_016134.3 | NP_057218.1 | ||
XM_017012953.1 | XP_016868442.1 | |||
XM_017012954.1 | XP_016868443.1 | |||
XM_017012955.1 | XP_016868444.1 | |||
AK075132.1 | BAC11423.1 | |||
AK296013.1 | ||||
AK304008.1 | ||||
AK315447.1 | ||||
BC020689.1 | AAH20689.1 | |||
DA699663.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576596 | Chr.8:96466318 - 96702414 on Build GRCh38 | Gain | CPQ SDC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |