Assay Details
Target Gene Details
Entrez Gene ID: | 245972 |
Gene Name: | ATPase H+ transporting V0 subunit d2 |
Gene Aliases: |
ATP6D2, VMA6 |
Location: |
Chr.8:86098910-86154225 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6V0D2 | NM_152565.1 | NP_689778.1 | ||
AK096027.1 | BAC04679.1 | |||
AY079172.1 | AAL87000.1 | |||
BC065207.1 | AAH65207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761443 | Chr.8:86097139 - 86158585 on Build GRCh38 | Gain |
![]() |
nsv611673 | Chr.8:86094481 - 86124091 on Build GRCh38 | Loss |
![]() |
nsv1028959 | Chr.8:85823138 - 86130154 on Build GRCh38 | Gain |
![]() ![]() |
dgv7262n100 | Chr.8:86099478 - 86165139 on Build GRCh38 | Gain |
![]() |
nsv428204 | Chr.8:85493249 - 86299979 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3891432 | Chr.8:85988884 - 86136762 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)