Assay Details
Target Gene Details
Entrez Gene ID: | 1978 |
Gene Name: | eukaryotic translation initiation factor 4E binding protein 1 |
Gene Aliases: |
4E-BP1, 4EBP1, BP-1, PHAS-I |
Location: |
Chr.8:38030502-38060365 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF4EBP1 | NM_004095.3 | NP_004086.1 | ||
AB044548.1 | BAB18650.1 | |||
AK312011.1 | ||||
BC004459.1 | AAH04459.1 | |||
BC058073.1 | AAH58073.1 | |||
BF964628.1 | ||||
BT007162.1 | AAP35826.1 | |||
CR456769.1 | CAG33050.1 | |||
L36055.1 | AAA62269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv610947 | Chr.8:37935047 - 38054626 on Build GRCh38 | Loss | ADRB3 GOT1L1 EIF4EBP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |