Assay Details
Target Gene Details
Entrez Gene ID: | 51435 |
Gene Name: | scavenger receptor class A member 3 |
Gene Aliases: |
APC7, CSR, CSR1, MSLR1, MSRL1 |
Location: |
Chr.8:27633665-27734027 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCARA3 | NM_016240.2 | NP_057324.2 | ||
NM_182826.1 | NP_878185.1 | |||
XM_017013535.1 | XP_016869024.1 | |||
XM_017013536.1 | XP_016869025.1 | |||
XM_017013537.1 | XP_016869026.1 | |||
AB007829.1 | BAA86201.1 | |||
AB007830.1 | BAA86202.1 | |||
BC060811.1 | AAH60811.1 | |||
BC075067.3 | AAH75067.1 | |||
BC075068.2 | AAH75068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv610867 | Chr.8:27607002 - 27757379 on Build GRCh38 | Gain | MIR6843 MIR3622B SCARA3 MIR3622A CLU CCDC25 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |