Assay Details
Target Gene Details
Entrez Gene ID: | 7201 |
Gene Name: | thyrotropin releasing hormone receptor |
Gene Aliases: |
TRH-R |
Location: |
Chr.8:109085745-109119785 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRHR | NM_003301.5 | NP_003292.1 | ||
XM_011517263.2 | XP_011515565.1 | |||
AY493373.1 | AAR84356.1 | |||
BC105045.1 | AAI05046.1 | |||
BC113360.1 | ||||
D16845.1 | BAA04120.1 | |||
X72089.1 | CAA50979.1 | |||
X75071.1 | CAA52965.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |