Assay Details
Target Gene Details
Entrez Gene ID: | 79648 |
Gene Name: | microcephalin 1 |
Gene Aliases: |
BRIT1, MCT |
Location: |
Chr.8:6406592-6648505 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv6868n100 | Chr.8:6266604 - 6447293 on Build GRCh38 | Gain | LOC100287015 MCPH1 |
esv3576515 | Chr.8:6377962 - 6478027 on Build GRCh38 | Gain | LOC100287015 MCPH1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |