Assay Details
Target Gene Details
Entrez Gene ID: | 4482 |
Gene Name: | methionine sulfoxide reductase A |
Gene Aliases: |
PMSR |
Location: |
Chr.8:10054224-10428891 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MSRA | NM_001135670.2 | NP_001129142.1 | ||
NM_012331.4 | NP_036463.1 | |||
XM_011543822.1 | XP_011542124.1 | |||
XM_011543823.2 | XP_011542125.1 | |||
XM_017013448.1 | XP_016868937.1 | |||
AF086925.1 | AAP97154.1 | |||
AF183420.1 | AAG09689.1 | |||
AJ242973.1 | CAB59628.1 | |||
AY958429.1 | AAY17426.1 | |||
BC054033.1 | AAH54033.1 | |||
CK819753.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |