Assay Details
Target Gene Details
Entrez Gene ID: | 51001 |
Gene Name: | mitochondrial transcription termination factor 3 |
Gene Aliases: |
CGI-12, MTERFD1 |
Location: |
Chr.8:96239398-96261613 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTERF3 | NM_001286643.1 | NP_001273572.1 | ||
NM_015942.4 | NP_057026.3 | |||
XM_006716570.3 | XP_006716633.1 | |||
XM_011517054.2 | XP_011515356.1 | |||
AF132946.1 | AAD27721.1 | |||
AK001801.1 | ||||
AK309002.1 | ||||
BC012995.2 | AAH12995.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6318 | Chr.8:96219001 - 96245773 on Build GRCh38 | Insertion | MTERF3 UQCRB |
nsv1020494 | Chr.8:96188447 - 96286981 on Build GRCh38 | Gain | MTERF3 PTDSS1 UQCRB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |