Assay Details
Target Gene Details
Entrez Gene ID: | 101929550 |
Gene Name: | uncharacterized LOC101929550 |
Gene Aliases: |
- |
Location: |
Chr.8:35672163-35710320 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929550 | NR_125819.1 | |||
AL137475.1 |
Target Gene Details
Entrez Gene ID: | 137970 |
Gene Name: | unc-5 netrin receptor D |
Gene Aliases: |
PRO34692, Unc5h4 |
Location: |
Chr.8:35235457-35796550 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UNC5D | NM_001322818.1 | NP_001309747.1 | ||
NM_080872.3 | NP_543148.2 | |||
XM_011544400.2 | XP_011542702.1 | |||
XM_011544401.2 | XP_011542703.1 | |||
XM_011544402.2 | XP_011542704.1 | |||
XM_011544403.2 | XP_011542705.1 | |||
XM_011544404.2 | XP_011542706.1 | |||
XM_017013044.1 | XP_016868533.1 | |||
XM_017013045.1 | XP_016868534.1 | |||
XM_017013046.1 | XP_016868535.1 | |||
XM_017013047.1 | XP_016868536.1 | |||
XM_017013048.1 | XP_016868537.1 | |||
XM_017013049.1 | XP_016868538.1 | |||
XM_017013050.1 | XP_016868539.1 | |||
AB055056.1 | BAB83663.1 | |||
AK122615.1 | ||||
AY358147.1 | AAQ88514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6152 | Chr.8:35678315 - 35711831 on Build GRCh38 | Insertion | UNC5D LOC101929550 |
nsv831285 | Chr.8:35628356 - 35790599 on Build GRCh38 | Loss | UNC5D LOC101929550 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |